Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XRCC1 (X-Ray Repair Cross Complementing Protein 1) Ab-1 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS434B0 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Conjugate:
Biotin
Unconjugated
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
Prediluted
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Form Concentration
MS434B0 100 μL Biotin Concentrated 200μg/mL
MS434P0 100 μL Unconjugated Concentrated 200μg/mL
MS434P1ABX 100 μL Unconjugated Concentrated 1mg/mL
MS434PABX 200 μL Unconjugated Concentrated 1mg/mL
MS434P1 500 μL Unconjugated Concentrated 200μg/mL
MS434R7 7 mL Unconjugated Prediluted Prediluted
MS434B1 500 μL Biotin Concentrated 200μg/mL
MS434B 1 mL Biotin Concentrated 200μg/mL
MS434P 1 mL Unconjugated Concentrated 200μg/mL
MS434R1 1 mL Unconjugated Prediluted Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. MS434B0 Supplier Epredia Supplier No. MS434B0

Provide accurate, reproducible results with the Epredia™ XRCC1 (X-Ray Repair Cross Complementing Protein 1) Ab-1, Mouse Monoclonal Antibody.

X-Ray Repair Cross Complementing (XRCC1) plays a role in excision repair of DNA after ionizing irradiation. XRCC1 binds to DNA ligase III through the C-terminal 96 amino acids and to DNA polymerase β through the N-terminal half. In testis XRCC1 is expressed at high levels. Cells with a mutation of this gene exhibit decreased single strand break repair and reduced recombination repair. They show increased double strand breaks, and sister chromatid exchange is increased up to 10-fold. The XRCC1 might serve as a scaffold protein during base excision-repair.

Host Species: Mouse

Clone: 33-2-5

Isotype: IgG2b

Species Reactivity: Human and Rat. Others not known.

Epitope: Not determined

Immunogen: Recombinant human XRCC1 protein

Molecular Weight: 85kDa

Positive Control: Ls174T, HT29 or HeLa cells; normal testis

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Native verified)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen XRCC1 (X-Ray Repair Cross Complementing Protein1) Ab-1
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 33-2-5
Concentration 200μg/mL
Conjugate Biotin
Host Species Mouse
Immunogen Recombinant human XRCC1 protein
Quantity 100 μL
Regulatory Status RUO
Research Discipline DNA Repair
Primary or Secondary Primary
Target Species Human, Rat
Form Concentrated
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.