Learn More
XRCC1 (X-Ray Repair Cross Complementing Protein 1) Ab-1 Mouse Monoclonal Antibody, Epredia™
Shop All Epredia ProductsDescription
Host Species: Mouse
Clone: 33-2-5
Isotype: IgG2b
Species Reactivity: Human and Rat. Others not known.
Epitope: Not determined
Immunogen: Recombinant human XRCC1 protein
Molecular Weight: 85kDa
Positive Control: Ls174T, HT29 or HeLa cells; normal testis
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunoprecipitation (Native verified)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | XRCC1 (X-Ray Repair Cross Complementing Protein1) Ab-1 |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | 33-2-5 |
| Concentration | Prediluted |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Recombinant human XRCC1 protein |
| Quantity | 7 mL |
| Regulatory Status | RUO |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.