Learn More
Labconco™ Atmospure Adapter Kit
Adapters allow 3/4 in. hose to connect to 3/8 in. fittings on glove boxes
$372.50
Specifications
Weight (English) | 5 lb. |
---|---|
Weight (Metric) | 2.3 Kg |
Product Type | Glove Box Kit |
Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
16-200-475
![]() |
Labconco™
5441901 |
Protector™ Glove Box |
Each for $372.50
|
|
|||||
16-200-474
![]() |
Labconco™
5441900 |
Precise™ Controlled Atmosphere Glove Box |
Each for $372.50
|
|
|||||
Description
AtmosPure 3/4 in. to 3/8 in. Adapter Kit has fittings that permit 3/4 in. ID neoprene hose included with 5441800 and 5441801 Kits (sold separately) to attach to glove box’s 3/8 in. inlet/outlets. Two fittings included with each kit. Elbow adapters for use with Labconco Precise Glove Boxes.Specifications
5 lb. | |
2.3 Kg | |
Glove Box Kit |