Learn More
Labconco™ Atmospure Cable
Oxygen/Moisture monitor cable for use with Labconco Protector Glove Boxes
$203.00 - $215.00
Specifications
| For Use With (Equipment) | AtmosPure |
|---|---|
| Weight (English) | 2 lb. |
| Weight (Metric) | 0.9 Kg |
| Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
16-200-476
![]() |
Labconco™
5442000 |
Atmospure 6-Pin Cable |
Each for $215.00
|
|
|||||
16-200-477
![]() |
Labconco™
5442100 |
Atmospure 9-Pin Cable |
Each for $203.00
|
|
|||||
Description
AtmosPure 9-Pin Oxygen/Moisture Monitor Cable is required for use when the AtmosPure is used in AUTO mode. Allows for automatic control of the AtmosPure blower from an accessory Oxygen or Moisture Monitor (sold separately).Specifications
| AtmosPure | |
| 2 lb. | |
| 0.9 Kg |