Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ CApture™ Portable Fuming System
SDP

Catalog No. 09682772 Shop All Labconco Corporation Products
Change view
Click to view available options
Voltage:
115 V
230 V
Amperage:
0.5 A
1 A
Plug Type:
China/Australia Plug
Domestic
GB Plug
Schuko Plug
Certifications/Compliance:
CE
ETL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Voltage Amperage Plug Type Certifications/Compliance
09-682-772 230 V 0.5 A Schuko Plug CE
09-682-770 115 V 1 A Domestic ETL
09-682-771 230 V 0.5 A GB Plug CE
09-682-773 230 V 0.5 A China/Australia Plug CE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Catalog No. 09-682-772 Supplier Labconco™ Supplier No. 3420022
Add to Cart
Add to Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.

Quickly fixes latent fingerprints at a crime scene using microburst or traditional CA fuming methods, while protecting the user from hazardous fumes

Unique design combines all the components needed in one compact system: a heater to heat the Super Glue™; a shatter-resistant, clear protective chamber to contain the fumes as they react with latent prints; an exhaust blower; and particulate/carbon exhaust filter. No ducting is required. Since prints may be easily developed at the scene, irrelevant evidence may be ruled out thus saving lab processing time and storage space.

  • Epoxy-coated steel housing with clear, shatter-resistant chamber
  • Leveling foot with locking pin stabilizes developer in upright position
  • Inverter ready, low energy usage (75w) allows system to run on car battery when connected to user-supplied inverter
  • Three-way heater/off/vent switch activates heater during print development and activates vent after prints have been fixed
  • Particulate/activated carbon filter cartridge traps smoke and CA fumes exhausted from chamber; cartridge has capacity for 100 cycles
  • Microprocessor-controlled heater allows user to set temperature in one-degree increments from 120° to 350°F (50° to 177°C)
  • Once set, temperature of heater stabilizes at set point (±15° F) within 3 to 5 minutes
  • Built-in blower pulls air from chamber, through filter and exhausts it back into room
  • Accessory Exhaust Connection Kit allows air to be exhausted out window or into fume hood
  • Instructions by video (available online) for setup and use

Compliance

CE conformity marking.

Specifications

Product Type Cyanoacralate Fuming Chamber
Coating Epoxy Powder
Filter Type Redundant HEPA and Bonded Carbon
Material Stainless Steel
Amperage 0.5 A
Certifications/Compliance CE
Depth (English) 10.75 in.
Depth (English) Interior 8 in.
Depth (Metric) 27.3 cm
Depth (Metric) Interior 20 cm
Dimensions (D x W x H) 8 x 8 x 16 in. (20 x 20 x 41 cm)
Height (English) 19 in.
Height (English) Interior 16 in.
Height (Metric) 48.26 cm
Height (Metric) Interior 41 cm
Plug Type Schuko Plug
Temperature Range (English) 120° to 350°F
Temperature Range (Metric) 50° to 177°C
Voltage 230 V
Weight (English) 30 lb.
Weight (Metric) 13.6 kg
Width (English) 15.5 in.
Width (English) Interior 8 in.
Width (Metric) 39.37 cm
Width (Metric) Interior 20 cm
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.