Learn More
Labconco™ Core Plate
SDP
Catalog No. 08640 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
Kjeldahl nitrogen apparatus
Catalog No. For Use With (Equipment)
08-640 Kjeldahl nitrogen apparatus
1 options
Catalog No. 08-640 Supplier Labconco™ Supplier No. 2011500
Add to Cart
Add to Cart
Core plate supports the heater coils for Labconco Macro Kjeldahl Apparatus.
Core plate supports the heater coils for Labconco Macro Kjeldahl Apparatus. Package of 6.
Specifications
Product Type Core Plate
For Use With (Equipment) Kjeldahl nitrogen apparatus
WARNING: Cancer - www.P65Warnings.ca.gov