Learn More
Labconco™ Ducting Parts: Connectors
SDP
Connectors for Labconco ducts
$679.50 - $853.50
Specifications
For Use With (Equipment) Hoods and Enclosures
View More Specs
Products 2
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
16-200-382
SDP
View Documents
Labconco™
5440900
Connector, Exhaust (Six Thimble)
Each for $853.50
Only null left
Add to Cart
16-200-383
SDP
View Documents
Labconco™
5440800
Connector, Hard Duct
Each for $679.50
Only null left
Add to Cart
Description
Description
Powder-coated steel, 6 in. diameter, for connecting the glove box to ductwork attached to a remote blower. Remote blower must be balanced with glove box's built-in blower.
Specifications
Specifications
Hoods and Enclosures
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.