Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ FilterMate™ Portable Exhauster: Gaseous Contaminants
SDP

Catalog No. 16304143 Shop All Labconco Corporation Products
Change view
Click to view available options
Voltage:
115 V
230 V
Hertz:
50 Hz
60 Hz
Dimensions (L x W x H):
14 x 23 x 22.7 in.
14.9 x 23 x 22.7 in.
Certifications/Compliance:
CAN/CSA C22.2, UL
CE
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Voltage Hertz Dimensions (L x W x H) Certifications/Compliance
16-304-143 230 V 50 Hz 14.9 x 23 x 22.7 in. CE
10-304-34 230 V 50 Hz 14 x 23 x 22.7 in. CE
16-304-142 115 V 60 Hz 14.9 x 23 x 22.7 in. CAN/CSA C22.2, UL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Catalog No. 16-304-143 Supplier Labconco™ Supplier No. 3970024
Add to Cart
Add to Cart
Request a Quote
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.

Includes

8 ft. of 5 in. I.D. gray flexible hose, two clamps and one 5 in. O.D. Upper Transition Adaptor for connection to Demonstration Hood, XPert Balance Enclosure, Protector XVS Ventilation Station or Protector Work Station.

Requires

Carbon Filter (Models for use with carbon filter only). Models for use with carbon filter require Organic Vapor Cell, Formaldehyde Impregnated Cell, or Ammonia Impregnated Cell. Combination models can support use of two carbon filters by removing HEPA filter.

Removehazardous powders, particulates or vapors from exhaust air stream, then return filtered air to environment by using a Labconco FilterMate™ Portable Exhauster for gaseous contaminants.

An accessory for Demonstration Hood, XPert Balance Enclosure, Protector XVS Ventilation Station or Protector Work Station.

  • Compact, corrosion-resistant housing of glacier white and light gray, dry powder, epoxy-coated steel with rubber feet
  • Uses HEPA filter, carbon-based filter or combination of both filters
  • Variable speed, non-sparking motorized impeller, adjustable to optimize airflows
  • Negative pressure plenum surrounds positive pressure impeller to capture and refilter unfiltered air
  • Front-mounted rocker switch controls power to blower
  • Rear-mounted auxiliary electrical outlet for connection to airflow monitor
  • Suitable for 2 ft., 3 ft. and 4 ft. wide x 22.75 in. high enclosures
  • Comes with IEC 3-wire electrical cord with plug

Models for use with HEPA filter

  • Include true bag-in bag-out filter disposal system for safely removing and replacing filter without detaching hose
  • Filter efficiency: 250 CFM
Models for use with Carbon-based filter
Filter efficiency: 250 CFM
Models for use with HEPA and Carbon-based filters
Filter efficiency: 200 CFM

Compliance

UL 3101-1/61010-1, CAN/CSA C22.2 No. 1010.1

Specifications

Product Type Exhauster
Filter Type Ammonia-Amine, Formaldehyde, Organic Vapor
Certifications/Compliance CE
Dimensions (L x W x H) 14.9 x 23 x 22.7 in.
Flow Rate 250 cfm
Height (English) 22.7 in.
Height (Metric) 58 cm
Hertz 50 Hz
Length (English) 14.9 in.
Length (Metric) 38 cm
Voltage 230 V
Weight (English) 91 lb.
Weight (Metric) 41 kg
Width (English) 1.2 ft.
Width (Metric) 38 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.