Learn More
Labconco™ FilterMate™ Portable Exhauster: Gaseous Contaminants



Removehazardous powders, particulates or vapors from exhaust air stream, then return filtered air to environment by using a Labconco FilterMate™ Portable Exhauster for gaseous contaminants.
$5190.00 - $5820.00
Specifications
Product Type | Exhauster |
---|---|
Flow Rate | 250 cfm |
Height (English) | 22.7 in. |
Height (Metric) | 58 cm |
Includes: 8 ft. of 5 in. I.D. gray flexible hose, two clamps and one 5 in. O.D. Upper Transition Adaptor for connection to Demonstration Hood, XPert Balance Enclosure, Protector XVS Ventilation Station or Protector Work Station.
Requires: Carbon Filter (Models for use with carbon filter only). Models for use with carbon filter require Organic Vapor Cell, Formaldehyde Impregnated Cell, or Ammonia Impregnated Cell. Combination models can support use of two carbon filters by removing HEPA filter.
Catalog Number | Mfr. No. | Certifications/Compliance | Hertz | Voltage | Dimensions (L x W x H) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Certifications/Compliance | Hertz | Voltage | Dimensions (L x W x H) | Price | Quantity & Availability | ||
10-304-34
![]() |
Labconco™
3970021 |
CE | 50 Hz | 230 V | 14 x 23 x 22.7 in. (36 x 58 x 58 cm) |
Each for $5,190.00
|
|
||
16-304-142
![]() ![]() |
Labconco™
3970004 |
CAN/CSA C22.2, UL | 14.9 x 23 x 22.7 in. (38 x 58 x 58 cm) |
Each for $5,820.00
|
|
||||
16-304-143
![]() |
Labconco™
3970024 |
CE | 14.9 x 23 x 22.7 in. (38 x 58 x 58 cm) |
Each for $5,650.00
|
|
||||
Description
An accessory for Demonstration Hood, XPert Balance Enclosure, Protector XVS Ventilation Station or Protector Work Station.
- Compact, corrosion-resistant housing of glacier white and light gray, dry powder, epoxy-coated steel with rubber feet
- Uses HEPA filter, carbon-based filter or combination of both filters
- Variable speed, non-sparking motorized impeller, adjustable to optimize airflows
- Negative pressure plenum surrounds positive pressure impeller to capture and refilter unfiltered air
- Front-mounted rocker switch controls power to blower
- Rear-mounted auxiliary electrical outlet for connection to airflow monitor
- Suitable for 2 ft., 3 ft. and 4 ft. wide x 22.75 in. high enclosures
- Comes with IEC 3-wire electrical cord with plug
Models for use with HEPA filter
- Include true bag-in bag-out filter disposal system for safely removing and replacing filter without detaching hose
- Filter efficiency: 250 CFM
Filter efficiency: 250 CFM
Models for use with HEPA and Carbon-based filters
Filter efficiency: 200 CFM
Compliance
UL 3101-1/61010-1, CAN/CSA C22.2 No. 1010.1
Specifications
Exhauster | |
250 cfm | |
22.7 in. | |
58 cm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.