Learn More
Labconco™ FreeZone™ Freeze-Dry Glass Trap
SDP
Catalog No. 09039070 Shop All Labconco Corporation Products
Change view
Click to view available options
Material:
Glass
Catalog No. Material
09-039-070 Glass
1 options
Catalog No. 09-039-070 Supplier Labconco™ Supplier No. 7871500
Add to Cart
Add to Cart
For use in FreeZone freeze dry units
  • Replacement glass trap
  • Fits inside CentriVap Cold Trap collector chamber
  • Protects chamber and lid from corrosive materials
  • Includes insulative cover, 1/2 in. stems and elbows
  • Not compatible with pre-2005 CentriVap Cold Traps
Specifications
Material Glass
For Use With (Equipment) Freezone™ freeze dry units
Type Trap
WARNING: Reproductive Harm - www.P65Warnings.ca.gov