Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ Glove Box Filters
SDP

For Labconco Filtered Glove Boxes

Supplier:  Labconco™ 5065100

 View more versions of this product

Catalog No. 16-101-15


Only null left
Add to Cart

Description

Description

16-101-11

  • Carbon Exhaust
  • Use as optional second outlet filter for removal of gaseous organic contaminants in low level radioisotope or chemical carcinogen applications
16-101-15
  • HEPA
  • Inlet or exhaust
  • Use as replacement filter or optional second outlet filter for removal of 0.3micrometer particulate contaminants, For Labconco Multi-Hazard Glove Boxes

Specifications

Specifications

Multi Hazard Glove Box
6 lb.
2.7 Kg
Optional second outlet filter for removal of 0.3μm particulate contaminants
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.