Learn More
Labconco™ Inflow and Downflow Sensor Kit
SDP
Catalog No. 3405500 Shop All Labconco Corporation Products
Change view
Click to view available options
Sensor Types:
Airflow
Catalog No. Sensor Types
34-055-00 Airflow
1 options
Catalog No. 34-055-00 Supplier Labconco™ Supplier No. 3405500
Add to Cart
Add to Cart
Requires
Installation and certifier calibration
Allows display of inflow and downflow velocities on the LCD information center on MyLogic™ Digital Display.
    • Applicable Products: PuriCare Procedure Stations (2013 and newer), Purifier Axiom Biosafety Cabinets, Purifier Cell Logic and Cell Logic+ Biosafety Cabinets, Purifier Logic and Logic+ Biosafety Cabinets
    • Includes sensor, wiring harness and hardware
    • Not for use on Class II, Type B2 biosafety cabinets
Specifications
For Use With (Equipment) PuriCare Procedure Stations (2013 and newer), Purifier Axiom Biosafety Cabinets, Purifier Cell Logic and Cell Logic+ Biosafety Cabinets, Purifier Logic and Logic+ Biosafety Cabinets
Type Inflow and Downflow Sensor Kit
Indicator Displayed on Logic Screen
IP Rating None
Sensor Types Airflow
WARNING: Cancer - www.P65Warnings.ca.gov