Learn More
Labconco™ Paper Liners for Forensic Enclosures
SDP
Paper liners for use inside Protector Evidence Drying Cabinets.
$141.80 - $1050.00
Specifications
For Use With (Equipment) Protector Evidence Drying Cabinet
Product Type Liner
For Use With (Application) Collect Material that may Fall from Evidence
View More Specs
Products 6
Catalog Number Mfr. No. For Use With (Equipment) Dimensions Includes Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Dimensions Includes Price Quantity & Availability  
09-682-729
SDP
View Documents
Labconco™
3894200
Protector Evidence Drying Cabinet 34 x 27 x 2 in. 100 Sheets
Each for $556.00
Only null left
Add to Cart
09-682-728
SDP
View Documents
Labconco™
3869800
Protector Evidence Drying Cabinet 31 x 3 x 3 in. 25 Sheets
Each for $141.80
Only null left
Add to Cart
10-359-257
SDP
View Documents
Labconco™
3869804
Protector Evidence Drying Cabinet 13 x 11 x 7 in. 100 Sheets
Pack of 25 for $313.50
Only null left
Add to Cart
10-359-256
SDP
View Documents
Labconco™
3869802
Protector Evidence Drying Cabinet 13 x 11 x 7 in. 25 Sheets
Pack of 25 for $227.50
Only null left
Add to Cart
10-359-258
SDP
View Documents
Labconco™
3894201
Protector Evidence Drying Cabinet 13 x 11 x 7 in. 25 Sheets
Pack of 100 for $860.50
Only null left
Add to Cart
10-359-259
SDP
View Documents
Labconco™
3894202
Protector Evidence Drying Cabinet 13 x 11 x 7 in. 100 Sheets
Pack of 100 for $1,050.00
Only null left
Add to Cart
Description
Description
Paper Liners for use with Labconco Protector Evidence Drying Cabinets. White paper is pre-cut to fit the floor of the cabinet to collect material that may fall from evidence. Paper is polyethylene coated on one side.
Specifications
Specifications
Protector Evidence Drying Cabinet
Liner
Collect Material that may Fall from Evidence
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.