Learn More
Labconco™ Pressure Relief Valve Vent Kits for Precise™ Glove Boxes
SDP
Accessories for Precise Glove Boxes
Supplier:  Labconco™ 5241200
 View more versions of this product
Catalog No. 16-108-423
Only null left
Add to Cart
Description
Description
Pressure Relief Vent Kits provide the components necessary to exhaust the pressure relief valve during overpressurization. Require 5230600 Pressure Relief Valve (included with Precise Controlled Atmosphere Glove Boxes) and outside ducting. Include 3.9 in. OD x 3.8 in. ID thimble connection, gasket, two elbows, four clamps and 13 in. long tubing. Ductwork for outside venting is not included. Carbon-Filtered Pressure Relief Vent Kit includes activated carbon filter. For ducting to outside or room. May be used with 5238500 HEPA-Filtered Pressure Relief Vent Kit.
Specifications
Specifications
Precise Controlled Atmosphere Glove Box
2.3 Kg
Exhaust Pressure Relief Valve during Overpressurization
5 lb.
Carbon-filtered Pressure Relief Valve Vent Kit
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.