Description
Replacement filters for use with PuriCare Animal Research Stations.
Specifications
Specifications
| Filter Type | Charcoal |
| For Use With (Equipment) | 4 ft. PuriCare Procedure Station |
| Product Type | PuriCare Replacement Filter |
| For Use With (Application) | Adsorb Odors, Pollutants, Gases |