Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ Biosafety Cabinet/Procedure Station Accessory, Service Fixture Kit
GSA_VASDP

Catalog No. 1630531 Shop All Labconco Corporation Products
Change view
Click to view available options
Color:
Silver
Material:
Brass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Material
16-305-31 Silver Brass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-305-31 Supplier Labconco™ Supplier No. 3747500
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Hardware and instructions

Designed to work with PuriCare Procedure Stations and Purifier Logic and Logic + Biosafety Cabinets, this Labconco Service Fixture Kit makes required repairs easy when kept on hand.

  • Serrated hose tip valve with quarter-turn control handle for field installation onto left or right side of cabinet interior
  • Cabinets are factory-prepared to accommodate up to four fixtures

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Product Type Wall-mounted Cabinet
Color Silver
Material Brass
For Use With (Equipment) Purifier™ delta series cabinet
Includes Serrated hose tip valve with ball valve, hardware and instructions
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.