Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ Filters
SDP

Carbon-based and HEPA filters for use in Labconco enclosures which require periodic replacement

Supplier:  Labconco™ 3707901

 View more versions of this product

Catalog No. 16-304-68


Only null left
Add to Cart

Description

Description

Carbon-based and HEPA filters for use in Labconco enclosures which require periodic replacement

Replacement HEPA Filter, 99.99% efficient on particles 0.3 micron in size. Not suitable for use with biohazardous materials or volatile organic material. For use with Labconco Purifier Class I Enclosures, Purifier Filtered PCR Enclosures, Purifier HEPA Filtered Enclosures, Purifier Vertical Clean Benches, XPert Filtered Balance Stations, XPert and RXPert Filtered Balance Systems.
Specifications

Specifications

99.99%
3 and 6 ft. Purifier™ Class I Safety, HEPA Filtered, Vertical Clean Bench and PCR Enclosures
36.7 Kg
76.2 to 101.6 cm
Particulate
HEPA
81 lb.
30 to 40 in.
Filter
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.