Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ Purifier™ Logic+ Class II B2 Biosafety Cabinets, 6 ft. Width, 8 in. Opening, International Model
SDP

Catalog No. 303681020 Shop All Labconco Corporation Products
Change view
Click to view available options
Includes:
Base Stand
Base Stand, Service Fixtures, Pass-through
Cabinet Only
Service Fixtures, Pass-through
Lighting:
Fluorescent
Fluorescent, UV
Plug Type:
China, Australia (230 V)
India (230 V)
International
Schuko (230 V)
UK (230 V)
Amperage:
12 A
7 A
8 A
20 product options available for selection
Product selection table with 20 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes Lighting Plug Type Amperage
30-368-1020 Service Fixtures, Pass-through Fluorescent, UV International 12 A
30-368-0030 Cabinet Only Fluorescent Schuko (230 V) 8 A
30-368-0020 Cabinet Only Fluorescent International 12 A
30-368-0050 Cabinet Only Fluorescent UK (230 V) 8 A
30-368-0040 Cabinet Only Fluorescent China, Australia (230 V) 8 A
30-368-0070 Cabinet Only Fluorescent India (230 V) 7 A
30-368-0120 Base Stand Fluorescent International 12 A
30-368-0130 Base Stand Fluorescent Schuko (230 V) 8 A
30-368-0140 Base Stand Fluorescent China, Australia (230 V) 8 A
30-368-0150 Base Stand Fluorescent UK (230 V) 8 A
30-368-0170 Base Stand Fluorescent India (230 V) 7 A
30-368-1030 Service Fixtures, Pass-through Fluorescent, UV Schuko (230 V) 8 A
30-368-1040 Service Fixtures, Pass-through Fluorescent, UV China, Australia (230 V) 8 A
30-368-1070 Service Fixtures, Pass-through Fluorescent, UV India (230 V) 7 A
30-368-1050 Service Fixtures, Pass-through Fluorescent, UV UK (230 V) 8 A
30-368-1120 Base Stand, Service Fixtures, Pass-through Fluorescent, UV International 12 A
30-368-1130 Base Stand, Service Fixtures, Pass-through Fluorescent, UV Schuko (230 V) 8 A
30-368-1140 Base Stand, Service Fixtures, Pass-through Fluorescent, UV China, Australia (230 V) 8 A
30-368-1150 Base Stand, Service Fixtures, Pass-through Fluorescent, UV UK (230 V) 8 A
30-368-1170 Base Stand, Service Fixtures, Pass-through Fluorescent, UV India (230 V) 7 A
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 20 options available.
20 options
Catalog No. 30-368-1020 Supplier Labconco™ Supplier No. 303681020

Biohazard cabinets with advanced ergonomic design excel in user comfort and functionality

MyLogic™ operating system provides intuitive programming, color-coded menus, status messages and alarms with diagnostic troubleshooting — all using full sentences.

  • ECM blower technology combined with CAP (Constant Airflow Profile™) maintains precise volume of air as filters load
  • Interior-mounted LCD displays HEPA filter life remaining and cabinet status
  • Provides personnel, product and environmental protection; suitable for work in Biosafety Levels 1, 2 and 3
  • Cabinets are designed to be hard ducted to the outside
Inclination Technology™ Suite includes:
  • Flush-mounted stainless steel electrical receptacle covers with dampening hinges
  • Patented curved air grille with built-in armrest
  • Fully accessible controls and eye-level mounted color display with MyLogic operating system

Some models have base stands, UV lights, service fixtures, pass throughs and/or a combination of those features included. (Refer to nominal width, sash opening, electrical configuration and product specifications for details.)

Specifications

Certifications/Compliance ADA, NSF 49
Chamber Material Stainless Steel
Noise Level <63 dBA
Plug Type International
Height (English) 72.6 in.
Sash Opening (Metric) 20.3 cm
Display LCD
Housing Material Powder Coated Steel Exterior and Stainless Steel Interior
Includes Service Fixtures, Pass-through
Lighting Fluorescent, UV
Height (Metric) 184.4 cm
Width (English) 78.3 in.
Width (Metric) 198.8 cm
Height (English) Viewing 27 in.
Depth (English) 31.2 in.
Depth (Metric) 79.2 cm
Weight (English) 840 lb.
Weight (Metric) 381 Kg
Voltage 100 V
Amperage 12 A
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.