Learn More
Labconco™ Recirculation Kit for Protector™ Filtered and Combination Glove Boxes
Shop All Labconco Corporation Products
Description
Includes
Hose, clamps and powder-coated steel adapter
Contains the components necessary to connect the built-in blower exhaust to the inlet HEPA filter for recirculation of air, useful for dry atmosphere operations.
- Allows built-in blower exhaust to be connected to inlet HEPA/ULPA filter, providing total recirculation of air volume useful for semi-dry atmosphere operations
- Combine with other accessories such as Valves, Traps and Diaphragm Pumps, to maintain specific air chemistry conditions inside main chamber
- Shipping weight: 2.5kg (5.5 lb.)
Specifications
Specifications
| For Use With (Equipment) | Protector Filtered and Combination Glove Boxes |
| For Use With (Application) | Connects motor blower to inlet HEPA filter of glove box |