Learn More
Labconco™ Replacement Parts for Crude Fiber Apparatus
SDP
Catalog No. 096816 Shop All Labconco Corporation Products
Change view
Click to view available options
Includes:
Heat Control
Heating Element
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
09-681-6 Heating Element
09-681-9 Heat Control
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 09-681-6 Supplier Labconco™ Supplier No. 3003100
Add to Cart
Add to Cart
Graduated volumetric markings
  • Thick wall
  • For use with crude fiber apparatus
  • Borosilicate glass
Specifications
Product Type Heating Element
Display No Display
Amperage 3 A
Hertz 50/60 Hz
Voltage 115 V
Wattage 350 W
For Use With (Equipment) Crude fiber apparatus
Includes Heating Element
Material Borosilicate Glass
Quantity 1 Ea.