Learn More
Labconco™ Sash Stop Kit
Sash stop restricts how far a vertical-rising sash may be opened.
$75.90 - $91.70
| Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
09-039-052
![]() |
Labconco™
9724500P |
Protector Premier Hood, Protector XL Benchtop Hood |
Each for $75.90
|
|
|||||
10-369-203
![]() |
Labconco™
9410300 |
Protector Premier Hood, Protector XStream Hood |
Each for $91.70
|
|
|||||
Description
- For use with older models of Protector Premier, XL Benchtop, Stainless Steel Radioisotope and Stainless Steel Perchloric Acid Fume Hoods made before 2012.
- Restricts how far a vertical-rising sash may be opened.
- Plastic device may be easily field installed on the fixture panel of a Protector Hood, which have been factory-prepared to accommodate a sash stop at the 60% open position (18 in. to work surface).
- Alternate sash positions may be field drilled. Each Sash Stop Kit includes components for one sash.
- 1 kit required for 3 ft. to 8 ft. Premier, XL Benchtop, XStream, PVC, Stainless Steel and XL Floor-Mounted Fume Hoods with vertical-rising sashes.
- 2 kits required for 10 ft., 12 ft. XL Benchtop with dual sash, Pass-Through Fume Hoods.
Specifications
| Sash Stop |