Learn More
Labconco™ Soda Acid Trap
SDP
Catalog No. 1631614 Shop All Labconco Corporation Products
Change view
Click to view available options
Material:
Acrylonitrile
Catalog No. Material
16-316-14 Acrylonitrile
1 options
Catalog No. 16-316-14 Supplier Labconco™ Supplier No. 7772000
Secondary vacuum trap
Secondary trap connects in series with freeze dry system and vacuum pump to prevent migration of corrosive chemicals into pump interior.
  • Clear acrylonitrile body allows visual check of color indicating media
  • Media changes from white to blue when exhausted
  • 3/4 in. OD inlet and outlet connections
  • Includes 20 in. rubber tubing, 3/4 in.ID x 3/8 in. wall and one clamp
Specifications
Material Acrylonitrile
For Use With (Application) Prevent migration of corrosive chemicals into pump interior
For Use With (Equipment) Freeze dry system and vacuum pump
Includes 20 in. rubber tubing, 0.75 in. i.d. x 0.375 in. wall and one clamp, 0.75 in. o.d. inlet and outlet connections
Weight (English) 12 lb.
Weight (Metric) 5.4 kg