Learn More
Labconco™ SpillStopper™ Work Surfaces for 5ft. Protector™Premier™Hoods
GSA_VASDP
Catalog No. 10369159 Shop All Labconco Corporation Products
Change view
Click to view available options
Includes:
Left Rear
Left Rear, Right Rear
Left Side
Left Side, Right Rear
No Cutouts
Right Rear
Right Side
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
10-369-159 Left Rear, Right Rear
10-369-156 Right Rear
10-369-154 No Cutouts
10-369-155 Left Rear
10-369-157 Left Side
10-369-158 Right Side
10-369-160 Left Side, Right Rear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 10-369-159 Supplier Labconco™ Supplier No. 9500511
Add to Cart
Add to Cart
Keep your equipment safe from spills with the Labconco SpillStopper Work Surface Protector - specifically dished and contoured to conform to the interior liner of Protector Laboratory Hoods.
SpillStopper work surfaces are dished to contain spills
  • The front edge has large radius to aerodynamically direct airflow in hood
  • Some surfaces have cutouts for surface-mounted service fixture and/or 3 x 6 in. cup sink
  • Flat work surfaces may also be coupled with adjustable height base stand to provide auxiliary workspace
  • Service fixture and cup sink are sold separately
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
For Use With (Equipment) Protector XL Laboratory Hood
Includes Left Rear, Right Rear
Product Type Work Surface
Material Epoxy Resin
For Use With (Application) Laboratory Fume Hood Work
WARNING: Cancer - www.P65Warnings.ca.gov