Learn More
Labconco™ SpillStopper™ Work Surface for 8ft. Protector™Premier™and Protector™XL Hoods

Shop All Labconco Corporation Products


Description
SpillStopper work surfaces are dished to contain spills
- Molded from special formulation of corrosion-resistant epoxy resins
- Front edge has large radius to aerodynamically direct airflow into the fume hood
- Pre-drilled holes for venting and plumbing passthroughs are provided
- Some include 6 x 3 in. cupsink cutouts
9500820 and 9500802
Not for use with Protector Solvent Storage Cabinets
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| For Use With (Equipment) | Protector XL Laboratory Hood |
| Product Type | SpillStopper Work Surface (8 ft.) |
| For Use With (Application) | 8 Ft. Protector Premier and Protector XL Hoods |
| Cut Style | Left Side Cupsink Cutout |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.