Learn More
Labconco™ Vacuum Tubing Kit for CentriVap™ System and RapidVap™
SDP
Catalog No. 163167 Shop All Labconco Corporation Products
Change view
Click to view available options
Material:
Polyurethane
Catalog No. Material
16-316-7 Polyurethane
1 options
Catalog No. 16-316-7 Supplier Labconco™ Supplier No. 7814500
Add to Cart
Add to Cart
Vacuum Tubing Kit for use with Labconco CentriVap Vacuum Concentrators and RapidVap Evaporators.
Vacuum Tubing Kit for CentriVap Concentrators and RapidVap Evaporators includes two each 1/2 in. ID vacuum tubing, 48 in. length, and 4 tubing clamps.
Specifications
Material Polyurethane
For Use With (Equipment) CentriVap™ system
Type Vacuum Tubing Kit
Weight (English) 6 lb.
Weight (Metric) 2.7 kg
WARNING: Cancer - www.P65Warnings.ca.gov