Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ XPert™ Bulk Powder Enclosures
SDP

Catalog No. 10304161 Shop All Labconco Corporation Products
Change view
Click to view available options
Includes:
Center Access Opening
Left Access Opening
Right Access Opening
Width (Metric):
152.4 cm
182.8 cm
243.8 cm
Hertz:
50 Hz
60 Hz
Voltage:
115 V
230 V
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes Width (Metric) Hertz Voltage
10-304-161 Right Access Opening 152.4 cm 60 Hz 115 V
10-265-156 Left Access Opening 152.4 cm 60 Hz 115 V
10-265-158 Left Access Opening 152.4 cm 50 Hz 230 V
10-265-159 Center Access Opening 152.4 cm 50 Hz 230 V
10-265-161 Center Access Opening 182.8 cm 60 Hz 115 V
10-265-157 Center Access Opening 152.4 cm 60 Hz 115 V
10-265-160 Left Access Opening 182.8 cm 60 Hz 115 V
10-265-162 Left Access Opening 182.8 cm 50 Hz 230 V
10-265-163 Center Access Opening 182.8 cm 50 Hz 230 V
10-265-164 Left Access Opening 243.8 cm 60 Hz 115 V
10-265-165 Center Access Opening 243.8 cm 60 Hz 115 V
10-265-166 Left Access Opening 243.8 cm 50 Hz 230 V
10-265-167 Center Access Opening 243.8 cm 50 Hz 230 V
10-304-162 Right Access Opening 152.4 cm 50 Hz 230 V
10-304-165 Right Access Opening 243.8 cm 60 Hz 115 V
10-304-164 Right Access Opening 182.8 cm 50 Hz 230 V
10-304-166 Right Access Opening 243.8 cm 50 Hz 230 V
10-304-163 Right Access Opening 182.8 cm 60 Hz 115 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
Catalog No. 10-304-161 Supplier Labconco™ Supplier No. 3963502
Add to Cart
Add to Cart
Request a Quote

Requires

Venting; can be connected to a remote blower or house exhaust and exhausted to the outside.

Exhausts hazardous airborne particulates and fumes to outside

XPert Bulk Powder Enclosures require venting and can be connected to a remote blower or house exhaust and exhausted to the outside.

  • Clean-Sweep™ openings and a zone-perforated rear baffle enhance horizontal laminar airflow
  • With built-in Guardian™ 1000 digital airflow monitor
  • Upper dilution air supply with by-pass air
  • Upper containment sash foil and side-entry air foils
  • Stainless-steel work surface with front spill trough
  • Transfer caddy with caddy latch and positioning plate
  • Pushbutton operated electric hydraulic lift mechanism for raising the container to the optimal ergonomic height
  • 30 in. interior depth and 31.5 in. interior height
  • Tempered safety glass front sash and sides
  • 12 in. high sash opening; ergonomic angled sash with sealed gas-assisted springs hold sash open (28 in. loading height)
  • Prewired fluorescent light and switches for remote blower and light
  • Powder-coated steel exterior
  • 2 in. square-tube powder-coated steel base stand
  • Left-side mounted waste chute
  • Fourplex electrical receptacle and two utility ports

Order Info

For 115V 60Hz. International models are available; contact your Customer Service Representative.

Compliance

SEFA 1-2002, Modified ASHRAE 110-1995, ANSI Z9.5-1993, UL 3101-1/61010-1, CAN/CSA C22.2 No. 1010.1

Specifications

Product Type Bulk Powder Enclosure
Coating Epoxy
Lighting Type None
Material Stainless Steel
Certifications/Compliance SEFA 1-2002, ASHRAE 110-1995, ANSI Z9.5-1993, UL 3101-1/61010-1, CAN/CSA C22.2 No. 1010.1, CE
Depth (English) 37 in.
Depth (English) Interior 30 in.
Depth (Metric) 93.98 cm
Depth (Metric) Interior 76.2 cm
Height (English) 75.5 in.
Height (English) Interior 32 in.
Height (English) Sash Opening 12 in.
Height (Metric) 191.7 cm
Height (Metric) Sash Opening 30.5 cm
Hertz 60 Hz
Includes Right Access Opening
Ports 2 Utility
Voltage 115 V
Weight (English) 870 lb.
Width (English) 60 in.
Width (English) Interior 58.8 in.
Width (Metric) 152.4 cm
Width (Metric) Interior 149.4 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.