Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ XPert™ Bulk Powder Filtered Systems
SDP

Catalog No. 10265147 Shop All Labconco Corporation Products
Change view
Click to view available options
Includes:
Center Access Opening
Center Handle
Left Access Opening
Left Side handle
Right Access Opening
Right Side handle
Voltage:
115 V
230 V
Hertz:
50 Hz
50/60 Hz
60 Hz
Width (English):
60 in.
72 in.
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes Voltage Hertz Width (English)
10-265-147 Center Handle 230 V 50/60 Hz 72 in.
10-265-143 Center Handle 230 V 50/60 Hz 60 in.
10-265-144 Left Access Opening 115 V 60 Hz 72 in.
10-265-141 Center Access Opening 115 V 60 Hz 60 in.
10-265-140 Left Access Opening 115 V 60 Hz 60 in.
10-265-142 Left Side handle 230 V 50/60 Hz 60 in.
10-265-145 Center Access Opening 115 V 60 Hz 72 in.
10-265-146 Left Side handle 230 V 50/60 Hz 72 in.
10-304-153 Right Access Opening 115 V 60 Hz 60 in.
10-304-155 Right Access Opening 115 V 60 Hz 72 in.
10-304-156 Right Side handle 230 V 50 Hz 72 in.
10-304-154 Right Side handle 230 V 50/60 Hz 60 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Catalog No. 10-265-147 Supplier Labconco™ Supplier No. 3961621
Add to Cart
Add to Cart
Request a Quote
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.

Protects from hazardous airborne particulates and fumes generated during powder transfer operations

  • Clean-Sweep™ openings and a zone-perforated rear baffle enhance horizontal laminar airflow
  • With built-in Guardian™ 1000 digital airflow monitor
  • Upper dilution air supply with by-pass air
  • Upper containment sash foil and side-entry air foils
  • Stainless steel work surface with front spill trough
  • Transfer caddy with caddy latch and positioning plate
  • Push-button operated electric hydraulic lift mechanism for raising the container to the optimal ergonomic height
  • 30 in. interior depth and 32 in. interior height
  • Tempered safety glass front sash and sides
  • 12 in. high sash opening; ergonomic angled sash with sealed gas-assisted springs hold sash open (28 in. loading height)
  • Built-in blower; outside ducting not required
  • 99.99% efficient HEPA filter
  • True bag-in/bag-out filter disposal system
  • Pre-wired fluorescent light and switches for remote blower and light
  • Epoxy-coated steel exterior
  • 2 in. square-tube epoxy-coated steel base stand
  • Left-side mounted waste chute
  • Fourplex electrical receptacle and two utility ports

Compliance

SEFA 1-2002, Modified ASHRAE™ 110-1995, ANSI™ Z9.5-1993, UL 3101-1/61010-1, CAN/CSA C22.2 No. 1010.1

Specifications

Product Type Bulk Powder Enclosure
Coating Epoxy
Lighting Type Fluorescent
Material Stainless Steel
Certifications/Compliance SEFA 1-2002, ASHRAE 110-1995, ANSI Z9.5-1993, UL 3101-1/61010-1, CAN/CSA C22.2 No. 1010.1, CE
Depth (English) 37 in.
Depth (English) Interior 30 in.
Depth (Metric) 93.98 cm
Depth (Metric) Interior 76.2 cm
Height (English) 83 in.
Height (English) Interior 32 in.
Height (English) Sash Opening 12 in.
Height (Metric) 210.8 cm
Height (Metric) Sash Opening 30.5 cm
Hertz 50/60 Hz
Includes Center Handle
Ports 2 Utility
Voltage 230 V
Weight (English) 1105 lb.
Width (English) 72 in.
Width (Metric) 182.8 cm
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.