Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Hazardous Material Label: ETHYL ACETATE

Catalog No. 19107658
Click to view available options
Display Text:
ETHYL ACETATE

Provide workers with all of the chemical information needed for hazardous materials they may encounter. Brady™ Hazardous Material Labels are made of vinyl and display text in black on yellow.

  • Provides workers with chemical information of hazardous materials
  • Made from B-928 vinyl material
  • Label Size: W x H: 4.5 x 3.75 in.

Specifications

Adhesive Type Acrylic, Permanent, Cold-temperature
Color Black, Yellow
Material Vinyl (B-928)
Product Type Hazardous Materials Label
Display Text ETHYL ACETATE
Quantity 25/Pk.
Shape Rectangle
Size (English) 4.5 x 3.75 in. (W x H)
Thickness (English) 0.0049 in.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.