Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mirus Bio™ Label IT™ Nucleic Acid Labeling Kit, DNP

Catalog No. MIR3800
Click to view available options
Quantity:
100 μg
25 μg
This item is not returnable. View return policy
This item is not returnable. View return policy

Label IT™ Nucleic Acid Labeling Reagents for efficient, direct, non-enzymatic labeling of DNA and RNA.

  • Label Any DNA or RNA Template – Suitable for a wide range of applications
  • One-step Chemical Method – Easily and consistently control the labeling reactions
  • Adjustable Labeling Density – Achieve high sensitivity with optimally labeled DNA and RNA
  • Covalent Mechanism – Permanent, non-destructive modification of nucleic acid residues is ideal for many diverse applications; labels do not impact hybridization performance

NOTE: All the Label IT Reagents use the same technology, but each attach different fluorophores with distinct excitation and emission spectra.

Specifications

Detection Method Primer-probe Detection
Label or Dye Not Applicable
Final Product Type DNA (Labeled), RNA (Labeled)
Sample Type DNA/RNA
Labeling Target DNA/RNA
Product Type Label Reagent
Quantity 100 μg
Form Lyophilized
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.