Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mirus Bio™ Label IT™ Nucleic Acid Labeling Kit, DNP
Click to view available options
Quantity:
100 μg
25 μg
Description
- Label Any DNA or RNA Template – Suitable for a wide range of applications
- One-step Chemical Method – Easily and consistently control the labeling reactions
- Adjustable Labeling Density – Achieve high sensitivity with optimally labeled DNA and RNA
- Covalent Mechanism – Permanent, non-destructive modification of nucleic acid residues is ideal for many diverse applications; labels do not impact hybridization performance
NOTE: All the Label IT Reagents use the same technology, but each attach different fluorophores with distinct excitation and emission spectra.
Specifications
Specifications
Detection Method | Primer-probe Detection |
Label or Dye | Not Applicable |
Final Product Type | DNA (Labeled), RNA (Labeled) |
Sample Type | DNA/RNA |
Labeling Target | DNA/RNA |
Product Type | Label Reagent |
Quantity | 100 μg |
Form | Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction