Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mirus Bio™ Label IT™ Nucleic Acid Labeling Kit, MFP488

Catalog No. MIR7100 Shop All Mirus Bio LLC Products
Change view
Click to view available options
For Use With (Application):
Labels 100 μg nucleic acid
Labels 25 μg nucleic acid
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
MIR7100 Labels 100 μg nucleic acid
MIR7125 Labels 25 μg nucleic acid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MIR7100 Supplier Mirus Bio™ Supplier No. MIR 7100
Add to Cart
Add to Cart

Label IT™ Nucleic Acid Labeling Reagents for efficient, direct, non-enzymatic labeling of DNA and RNA.

  • Label Any DNA or RNA Template – Suitable for a wide range of applications
  • One-step Chemical Method – Easily and consistently control the labeling reactions
  • Adjustable Labeling Density – Achieve high sensitivity with optimally labeled DNA and RNA
  • Covalent Mechanism – Permanent, non-destructive modification of nucleic acid residues is ideal for many diverse applications; labels do not impact hybridization performance

NOTE: All the Label IT Reagents use the same technology, but each attach different fluorophores with distinct excitation and emission spectra.

  • MFP488 Nucleic Acid Labeling Reagent
  • High Photostability – Facilitates longer time for observation and image capture
  • Widespread Instrument Compatibility – Spectrally similar to fluorescein
  • Stable at Low pH – Fluorescence is maintained over a broad range (pH 4-9)

Specifications

Product Type Label Reagent
Concentration 1X
Product Line Molecular Probes, Click-iT
For Use With (Application) Labels 100 μg nucleic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.