Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mirus Bio Label IT™ siRNA Tracker™ Intracellular Localization Kit, CX-Rhodamine

Catalog No. MIR7214
Click to view available options
:
Intracellular Localization Kit, CX-Rhodamine
This item is not returnable. View return policy
This item is not returnable. View return policy

Label IT™ siRNA Tracker Intracellular Localization Kits is ideal for efficient, direct labeling for in vitro and in vivo tracking of siRNA.

  • Versatile Labeling: Efficiently label and visualize the siRNA of your choice
  • One-step Chemical Method: Easily and consistently control the labeling reactions and labeling density
  • High Efficiency Labeling: Optimal visualization of siRNA in cells and tissues

Specifications

Detection Method Fluorescence
Format Kit
Final Product Type DNA (Labeled), RNA (Labeled)
Labeling Target DNA/RNA
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.