Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mirus Bio Label IT™ Tracker™ Intracellular Nucleic Acid Localization Kit, CX-Rhodamine

Catalog No. MIR7022 Shop All Mirus Bio LLC Products
Change view
Click to view available options
Intracellular Nucleic Acid Localization Kit, CX-Rhodamine
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MIR7022 Intracellular Nucleic Acid Localization Kit, CX-Rhodamine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MIR7022 Supplier Mirus Bio Supplier No. MIR 7022

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Label IT Tracker Intracellular Nucleic Acid Localization Kit is ideal for efficient, direct labeling of plasmid DNA for in vitro and in vivo tracking experiments.

  • Versatile Labeling: Efficiently label and visualize the DNA of your choice
  • One-step Chemical Method: Easily and consistently control the labeling reactions
  • High Efficiency Labeling: Optimal visualization of DNA in cells
  • Superior Tracking: Monitor both subcellular localization and reporter transgene expression following transfection of labeled (plasmid) DNA

Specifications

Detection Method Fluorescence
Format Kit
Final Product Type DNA (Labeled), RNA (Labeled)
Labeling Target DNA/RNA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.