Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Multicolor Label Printer with WIFI and Bluetooth

Catalog No. 17100953
Click to view available options
Color:
Blue, Gray

Professional grade, premium experience. Print color visuals, cut shapes and let users walk up and print with the BradyPrinter Multicolor & Cut Sign and Label Printer.

Professional grade. Premium experience. Print color visuals, cut shapes and let users walk up and print with the BradyPrinter S3700 Multicolor and Cut Sign and Label Printer.

  • Put color at the core of your labeling for visuals that truly pop, including safety labels, lean visuals, lockout tags and work-in-process labels.
  • Create shapes, arrows, letters and more with the built-in XY plotter cutter that slices straight lines, smoother curves and tighter connection points.
  • Empower any user to print how they choose with time-saving onboard templates, intuitive PC labeling software and a mobile app.
  • Simply tap, print, go with preprogrammed onboard label templates plus 600 barcodes and symbols.
  • Get your print jobs done right in less time with 20-second ribbon and material changeovers plus up to 5-in.-per-second print speed.
  • Stack pipe markers, pack multiple shapes per label and enjoy printing right on the first label to minimize waste.
  • Optimized for 1-, 2- or4-color panel ribbons for multicolor made easy -- no calibrating, no threading, no waste.
  • Print ribbon lasts up to 60% longer by operating independently of label roll to waste less, print more.

Specifications

Product Type Label Printer
Color Blue, Gray
Display Type Color LCD Touchscreen
For Use With (Equipment) Brady Labels
Material Plastic
Print Technology Thermal Transfer
Length (English) 14.25 in.
Width (English) 19 in.
Height (English) 11 in.
Color Capability Multi-Color
Description S3700 Multicolor Label Printer
Display Size 6.11 x 3.42 in.
Includes (1) S3700 Safety and Facility Printer;(1) Brady Workstation Safety and Facility ID Software (BWS-SFIDS-NA-EM);(1) Quick Start Guide;(1) Power Cord;(1) USB Cord;(1) Mouse;(1) Cleaning Kit (PCK-6);(1) USB Flash Drive
Languages English (US);French (France);Portuguese (Portugal);Spanish (Spain);Bahasa (Indonesian);Bahasa Malay;Czech;Danish;German;Estonian;Croatian;Italian;Hungarian;Dutch;Norwegian;Polish;Romanian;Slovak;Slovenian;Finnish;Swedish;Vietnamese;Turkish;Bulgarian;Russian;Korean;Chinese (Traditional and Simplified);Thai;Japanese
Memory 512 MB
Model S3700
Print Speed 5 in./s
Resolution 300 dpi
Connector Types Bluetooth 5 Low Energy (class II)
Weight (English) 36.9 lb.
Certifications/Compliance cULus, CE, UL-AR & S Mark, UL NOM CoC, RCM Mark, EAC, KC, CCC, RoHS, BIS, BSMI, LOA, SABS
For Use With (Application) Asset Tracking;Cold temperature Labeling;Component and Equipment Labeling;Electrical Labeling;Facility Identification;General Identification;Hazard Communication Identification;Lean and 5S Labeling;Pipe and Valve Marking;Safety Identification;Security Identification;Temperature Indicating Labeling;Barcode Labeling;Exit and Egress Marking;Floor Marking;Food and Beverage Process Labeling;Healthcare and Hospital Labeling;Inventory and Inspection Labeling;Temporary Labeling;Warehouse Marking
Warranty 5 Years
Power Supply Details AC Adaptor
Voltage 110/240 V
Keyboard Type QWERTY (US)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.