Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STERIS LabKlenz™ 100 Alkaline Detergent
Shop All STERIS Corporation Products
Click to view available options
Volume (English):
1 gal.
5 gal.
30 gal.
250 gal.
Container Type:
Drum
Plastic Bottles
Plastic jerrican
Tote
Description
- Formulated to meet unique cleaning and materials compatibility requirements found in laboratory glassware washing applications
- High alkaline for effective removal of residues in both soft and hard water
- Contains corrosion inhibitors for excellent materials compatibility
- Reduces utility consumption and cleaning time with low foaming, free-rinsing formulation
- Phosphate-free formulation with biodegradable surfactants
- Manufactured in cGMP compliant, FDA registered and ISO 9001 certified facility
- Specific gravity: 1.15
Specifications
Specifications
| Sterility | Non-sterile |
| Appearance | Yellow |
| Certifications/Compliance | Certificate of analysis |
| Container Type | Tote |
| For Use With (Equipment) | Laboratory washers, IBS system |
| Volume (English) | 250 gal. |
| Volume (Metric) | 946.35 L |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction