Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STERIS LabKlenz™ 100 Alkaline Detergent

Catalog No. p-7231486
Click to view available options
Volume (English):
1 gal.
5 gal.
30 gal.
250 gal.
Container Type:
Drum
Plastic Bottles
Plastic jerrican
Tote

Alkaline detergent with corrosion inhibitors

  • Formulated to meet unique cleaning and materials compatibility requirements found in laboratory glassware washing applications
  • High alkaline for effective removal of residues in both soft and hard water
  • Contains corrosion inhibitors for excellent materials compatibility
  • Reduces utility consumption and cleaning time with low foaming, free-rinsing formulation
  • Phosphate-free formulation with biodegradable surfactants
  • Manufactured in cGMP compliant, FDA registered and ISO 9001 certified facility
  • Specific gravity: 1.15

Specifications

Appearance Yellow
Container Type Plastic Bottles
Quantity 4 Pk.
Volume (English) 1 gal.
Volume (Metric) 3.785 L
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.