Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STERIS™ LabKlenz 110 Alkaline Detergent

Catalog No. p-200076293 Shop All STERIS Corporation Products
Click to view available options
Size:
1 gal.
5 gal.
30 gal.

Catalog No. 17-467-596

Supplier: STERIS™ 1L1108

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

LabKlenz 110 Alkaline Detergent is a high alkaline detergent for difficult organic residues. This phosphate-free, liquid, alkaline detergent effectively removes residues in both soft and hard water. It reduces utility consumption and cleaning time with its low-foaming, free-rinsing formulation. pH (1%): 12.4, typical.

LabKlenz 110 Detergent is specially formulated to meet the unique cleaning and materials compatibility requirements found in laboratory glassware washing applications.

HOW LABKLENZ 110 DETERGENT WORKS

LabKlenz 110 Detergent is a phosphate-free, liquid, alkaline detergent for use in automated laboratory glassware washing applications. It is extremely free rinsing to meet critical laboratory requirements.

WHY LABKLENZ 110 DETERGENT?

  • Flexibility - Highly alkaline for effective removal of residues in both soft and hard water
  • Cost Savings - Reduces utility consumption and cleaning time due with low foaming, free-rinsing formulation
  • Environmentally Friendly - phosphate-free formulation with biodegradable surfactants
  • Reliability - Manufactured in a cGMP compliant, FDA registered and ISO 9001 certified facility
pH (1%): 12.4, typical

Specifications

Product Type Detergent
Color-coded No
Form Clear Liquid
pH 12.4
Scent Unscented
Sterility Non-sterile
Size 1 gal.
Volume (Metric) 3.78 L
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.