Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Laboratory Performance Check Solution (For EPA 531.1)

Catalog No. usppm5311
Encompass
Click to view available options
Quantity:
1 x 1 mL

Chemical Identifiers

CAS 1634-04-4
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
InChI Key BZLVMXJERCGZMT-UHFFFAOYSA-N
PubChem CID 15413
ChEBI CHEBI:27642
IUPAC Name 2-methoxy-2-methylpropane
SMILES CC(C)(C)OC

Specifications

Packaging Ampule
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) Each component in Methanol:
Aldicarb Sulfoxide    •100μg/mL
3-Hydroxycarbofuran    •2μg/mL
Methiocarb    •20μg/mL
BDMC    •10μg/mL
Chemical Name or Material Laboratory Performance Check Solution
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.