Learn More
Eisco™ Labradorite Mineral Specimen
SCI_ED
Catalog No. S26660 Shop All Eisco Products
Change view
Click to view available options
Quantity:
10/Pk.
Catalog No. Quantity
S26660 10/Pk.
1 options
Catalog No. S26660 Supplier Eisco™ Supplier No. ESNG0071PK10
Add to Cart
Add to Cart
Labradorite is a variety of plagioclase feldspar, an intermediate between anorthite and albite.
  • Exhibits iridescence in some samples, producing a brilliant array of colors
  • Correlates well to Next Generation Science Standards Evidence Statements
  • Chemically intermediate between albite and anorthite
  • Samples with clean crystal faces showcase iridescence
  • Ideal for geology classrooms and labs studying minerals
Specifications
Product Type Metamorphic Rock
Quantity 10/Pk.
Specimen Size 2 to 3 cm
Weight (Metric) 0.2 kg
Specimen Type Labradorite