Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Labtainer™ 2D BioProcess Container (BPC), three ports, MPCs, quick-connect
GREENER_CHOICE

Catalog No. p-7220327
Click to view available options
Capacity (Metric):
2 L
5 L
10 L
20 L
50 L

Thermo Scientific Labtainer 2D BioProcess Containers (BPCs) are composed of Aegis5-14 single-web, multi-layer film.

Features of the Labtainer 2D BPC include:
• Helps eliminate post-use cleaning steps required with re-useable containers and reduce cross-contamination risks
• Constructed in an ISO 7 cleanroom under cGMP conditions

Applications of the Labtainer 2D BPC include:
• Harvest from, feed into, and collect samples from bioreactors and fermentors
• Prepare and store buffer, culture media, and process liquid
• Collect chromatography feed and fractions
• Harvest, storage, and transport bulk drug product and bulk drug precursors

Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library with over 20,000 unique components that include:
• Fittings—straight, reducing, elbow, T-style, X-style, and cross-style
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors
• Filters—used for bioburden reduction; sterilization grade filters and vent filters
• Tubing—thermoplastic elastomers, platinum-cured silicone
• Sample ports—septum-style and needle-free ports
• Clamps—sliders, pinch, and crimp bands

TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Not Autoclavable
Capacity (Metric) 5 L
Certifications/Compliance ISO 7
Color Transparent
Disposable Single-use
For Use With (Application) Bioprocessing
Sterility Sterile
Tubing Style C-Flex
Film Description Aegis 5-14
Port Detail 3 ports
Quantity 5 L
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.