Learn More
Thermo Scientific™ 2D Labtainer™ BioProcess Container (BPC), ASI™ 28 film, 3 ports, Luer Lock connectors, injection site

Use small volume Labtainers for sampling, harvesting, preparation, storage and transport of process liquids.
$1862.00 - $2129.00
Specifications
Content And Storage | Store at room temperature. |
---|---|
Includes | 3 Ports (1 Luer Lock Insert Connection + Cap, 1 Luer Lock Body Connection + Cap, 1 Injection Site) |
Material | EVA |
Tubing Style | EVA |
Film Description | ASI™ 28 |
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
SS00006I
![]() |
Thermo Scientific™
SS00006I |
100 mL |
Case of 50 for $1,862.00
|
|
|||||
SS00007I
![]() |
Thermo Scientific™
SS00007I |
250 mL |
Case of 50 for $1,927.00
|
|
|||||
SS00008I
![]() |
Thermo Scientific™
SS00008I |
500 mL |
Case of 50 for $1,885.00
|
|
|||||
SS00009I
![]() |
Thermo Scientific™
SS00009I |
1 L |
Case of 50 for $2,061.00
|
|
|||||
SS00010I
![]() |
Thermo Scientific™
SS00010I |
2 L |
Case of 50 for $2,129.00
|
|
|||||
Description
This Thermo Scientific™ Labtainer™ BioProcess Container (BPC) 2D system is composed of Thermo Scientific™ ASI™ 28 film, a 4-layer, 12.5 mL, animal origin–free ethyl vinyl acetate film and includes 3 ports (including an injection site). A wide range of capacities, port configurations and tubing styles are available.
- Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
- All Labtainers constructed in an ISO 7 cleanroom environment under cGMP conditions
Applications:
- Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
- Preparing and storing buffer, culture media, and process liquid
- Collecting chromatography feed and fraction
- Harvesting, storing, and transporting bulk drug product and drug precursors
Specifications
Store at room temperature. | |
EVA | |
ASI™ 28 | |
Luer lock connectors, reducer and injection site | |
50/Cs. | |
Irradiated |
3 Ports (1 Luer Lock Insert Connection + Cap, 1 Luer Lock Body Connection + Cap, 1 Injection Site) | |
EVA | |
One 4-mil. poly bag | |
Labtainer™ | |
Room Temperature | |
Bioprocess Container |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.