Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ 2D Labtainer™ BioProcess Container (BPC), ASI™ 28 film, 3 ports, MPC insert and body connectors, injection site
GREENER_CHOICE

Use small volume Labtainers for sampling, harvesting, preparation, storage and transport of process liquids.

$1425.00 - $1734.00

Specifications

Content And Storage Store at room temperature.
Material EVA
Tubing Style EVA
Film Description ASI™ 28
Outer Packaging One 4-mil. poly bag
View More Specs

Products 4
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
SS00011I
GREENER_CHOICE
View Documents
Thermo Scientific™
SS00011I
100 mL
Case of 50 for $1,425.00
Only null left
Add to Cart
 
SS00012I
GREENER_CHOICE
View Documents
Thermo Scientific™
SS00012I
250 mL
Case of 50 for $1,509.00
Only null left
Add to Cart
 
SS00013I
GREENER_CHOICE
View Documents
Thermo Scientific™
SS00013I
500 mL
Case of 50 for $1,536.00
Only null left
Add to Cart
 
SS00014I
GREENER_CHOICE
View Documents
Thermo Scientific™
SS00014I
1 L
Case of 50 for $1,734.00
Only null left
Add to Cart
 
Description

Description

This Thermo Scientific™ Labtainer™ BioProcess Container (BPC) 2D system is composed of Thermo Scientific™ ASI™ 28 film, a 4-layer, 12.5 mL, animal origin–free ethyl vinyl acetate film and includes 3 ports (including an injection site). A wide range of capacities, port configurations and tubing styles are available.

  • Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
  • All Labtainers constructed in an ISO 7 cleanroom environment under cGMP conditions

Applications:

  • Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
  • Preparing and storing buffer, culture media, and process liquid
  • Collecting chromatography feed and fraction
  • Harvesting, storing, and transporting bulk drug product and drug precursors
Specifications

Specifications

Store at room temperature.
EVA
One 4-mil. poly bag
Labtainer™
Irradiated
EVA
ASI™ 28
MPC insert and body connectors and injection site
Room Temperature
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.