Learn More
Thermo Scientific™ 2D Labtainer™ BioProcess Container (BPC), ASI™ 26/77 film, 3 ports, MPC insert and body connectors

Description
This Thermo Scientific™ BioProcess Container (BPC) 2D System is composed of Thermo Scientific™ ASI™ 26/77 Polyethylene Film, a single-web, multi-layer 12.5 mL film. This BPC includes 2 ports (1 MPC insert + cap, 1 MPC body + cap). It is available in 50, 100 and 200L capacities.
- Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
- All Labtainers constructed in an ISO 7 cleanroom environment under cGMP conditions
Applications:
- Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
- Preparing and storing buffer, culture media, and process liquid
- Collecting chromatography feed and fraction
- Harvesting, storing, and transporting bulk drug product and drug precursors
BPC port specifications:
- Line 1: MPC insert with 9.5mm (3/8 in.) barb, cap, and pinch clamp. Tubing: Silicone, length: 51cm (20 in.), ID x OD: 9.5 x 15.9mm (0.38 x 0.63 in.)
- Line 2: MPC body with 9.5mm (3/8 in.) barb, plug, and pinch clamp.
Specifications
Specifications
| Content And Storage | Store at room temperature. |
| For Use With (Equipment) | TK-0500-05 |
| Includes | 2 Ports (1 MPC Insert + Cap, 1 MPC Body + Cap) |
| Material | Polyethylene |
| Tubing Style | C-Flex, 51 cm (20 in.) |
| Film Description | ASI™ 26/77 |
| Outer Packaging | 4 mil. poly bag, One 4-mil. poly bag |
| Port Detail | MPC connections + caps, pinch clamps |
| Product Line | Labtainer |
| Protective Inner Packaging | Poly bags on tubing ends, protective material around rough areas |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.