Learn More
Thermo Scientific™ Labtainer™ BioProcess Container (BPC), 10L with 3 Ports, MPC Insert, 2D BPC

Description
Using Thermo Scientific™ Labtainer™ BPC bag with 3 ports (MPC insert), simplify cleaning and disposal and reduce cross-contamination risks for 2 to 50L volumes of media, buffers, water for injection (WFI) and biological products.
Labtainers are constructed with our CX5-14 single web, multi-layer cast film manufactured in a cGMP facility. Our standard Labtainers are suitable for a wide array of bioprocess applications. For optimal fit, form and function, Labtainers can be customized using the industry's largest component library.
Highlights:
- 11 sizes from 50mL to 50L
- Film designed to incorporate best combination of flexibility and durability for collection, storage and transport of sterile bioprocess liquids: superior stress resistance, puncture resistance and physical properties to ensure stability of contents
- Gamma-irradiation sterilized
- Convenient end-port design with a handle at the opposing end
- Space-efficient, ergonomic alternative to rigid walled carboys
- Customizable via the industry's largest library of qualified components
Recommended for:
Sampling – collection of in-process samples at a variety of bioprocessing stages
Harvest – collection and storage of bulk product from bioreactors
Storage – sterile storage of media, buffers and process intermediates
Separation – feeding and receiving of sterile liquids from separation systems
Purification – buffer feeding to purification system, fraction collection and storage
Filling – sterile reservoir for filling systems
Transport & Shipping – bulk sterile liquids, buffers, media and process intermediates within and between facilities

Specifications
Specifications
Capacity (Metric) | 10 L |
Dimensions | 62.2 cm x 30 cm (24.5 in. x 11.8 in.) |
Outer Packaging | Packages placed in strong cardboard cartons with product and lot identification |
Protective Inner Packaging | Packed and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area |
Quantity | 10 L, each |
Sterilization Method | Gamma Irradiated |
Tubing Line 1 | Line 1. MPC insert, polycarbonate, Tubing: C-Flex, length: 30 cm (12 in.) ID x OD: 9.5 x 12.7 mm (0.38 x 0.5 in.) |
Tubing Line 3 | Line 3. Plugged, No Tubing |
Tubing Line 2 | Line 2. MPC insert, polycarbonate, Tubing: C-Flex, length: 30 cm (12 in.) ID x OD: 9.5 x 12.7mm (0.38 x 0.5 in.) |
Product Line | Labtainer™ |
Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.