Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Computype Labware Labeling Assessment
SDP

Catalog No. CMPLABLASMT
Change view
Click to view available options
Product Type:
Workplace Safety Compliance Assessments
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
CMPLABLASMT Workplace Safety Compliance Assessments
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
FDA 21CFR Approved materials FDA 21CFR UDI OSHA GHS 1910.1200 GMP compliance GLP compliance ANSI ISO barcode grading
Catalog No. CMPLABLASMT Supplier Computype Supplier No. CMPLABLASMT

To request a quote for this item, please select Request a Quote to reach our dedicated support team.

FDA 21CFR Approved materials FDA 21CFR UDI OSHA GHS 1910.1200 GMP compliance GLP compliance ANSI ISO barcode grading

A comprehensive evaluation of your current labeling processes designed to improve efficiency and reduce costs.

Smarter Labeling Starts Here - For over fifty years, Computype has partnered with life science and industrial organizations to solve complex labeling challenges in environments that demand precision, durability, and compliance. From barcoding in extreme conditions to meeting strict regulatory requirements, we specialize in keeping your samples, components, and products properly identified and traceable.

Whether you're just getting started, troubleshooting an existing system, or exploring more efficient solutions, we are here to help. Our process begins with understanding your goals and workflows - then we design a solution that fits, streamlines your operations, and ensures compliance every step of the way.

Let's simplify your labeling process without compromising reliability. Our labware labeling assessment reviews your current identification processes to uncover inefficiencies and recommend smarter strategies.We'll help streamline workflows, enhance traceability, reduce labeling costs, and solve problems.

  • Bar code printing, verification and scanning expertise that ensures performance and compliance.
  • On-demand and preprinted labeling options
  • Sequence management
  • Durable materials that withstand freezing, thawing, and chemical exposure to ensure your assets identification.
  • Cryogenic environments
  • Nase free environments
  • Extreme heat exposure
  • Sterilization ready materials
  • Enviromental friendly sustainable label materials
  • Label printer support and configuration optimization for your workflow requirements
  • Bar code label printing software and associated design label design programmin
  • Color coding for clarification and workflow efficiency
  • Automatic labeling automation
  • Labware Prep outsourcing the labeling of tubes, vials, microwell plates, etc. for ultimate convenience and accuracy
  • Bar coding of glass and plastic
  • Labware branding
  • Add a fill line or graduated line scale for improved accurary
  • Color coding
  • Tare weighing
  • Knowledge of various regulatory requirements covering FDA 21CFR materials, OSHA GHS, GLP, GMP, UDI, ISBT-128

Specifications

Product Type Workplace Safety Compliance Assessments
Federal Regulation OSHA 29 CFR, FDA 21 CFR
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.