Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mettler Toledo™ LabX™ Direct pH Software

Catalog No. 01915037 Shop All METTLER TOLEDO Products
Change view
Click to view available options
For Use With (Equipment):
pH Meters
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
01-915-037 pH Meters
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-915-037 Supplier Mettler Toledo™ Supplier No. 51302876

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

Allows an InMotion™ Autosampler to be connected to a SevenExcellence™ pH Meter. This combination maximizes throughput by allowing fully automated measurements of more than 300 samples.

  • Ensures that all data is stored centrally and securely, following requirements for data integrity according to ALCOA+
  • Allows an InMotion™ Autosampler to be connected to a SevenExcellence™ pH meter
  • Maximizes throughput by allowing fully automated measurements of up to 300 samples
  • The automated pH measurement system eliminates process-order and transcription errors without sacrificing bench space
  • Protects samples from environmental influence
  • Automation generates highly reproducible results
  • Ideally suited for transferring data to a PC
TRUSTED_SUSTAINABILITY

Specifications

Type Software
For Use With (Application) Fully automated measurements of more than 300 samples
For Use With (Equipment) pH Meters
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.