Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lac I Polyclonal Antibody, Rockland™
SDP

Catalog No. 501991966
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-1966 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-1966 Supplier Rockland Immunochemicals Supplier No. 600401B04
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This antibody reacts with recombinant Lac I protein. A BLAST analysis was used to suggest cross-reactivity with Lac I from DNA-binding transcriptional repressor in Escherichia coli (strain DH10B) and transcriptional repressor of the lac operon in Shigella sonnei (strain Ss046) based on a 100% homology with the immunizing sequence. Cross-reactivity with Lac I from other sources has not been determined.

The lac repressor protein (Lac I) is an important genetic control protein. It controls the expression of the lactose metabolizing enzymes in Escherichia coli. This regulation involves specific recognition of the operator DNA sequence (Lac O) and consequent inhibition of transcription initiation by RNA polymerase; modulation of binding at this target site by small sugar molecules, inducers, provides the basis for regulation. Different ligand binding activities are associated with the isolated domains of Lac I: specific and nonspecific DNA binding with the NH2 termini, and inducer and specific DNA binding with the core protein. This antibody could possibly be used to recover and purify protein/nucleic acid complexes (host and viral proteins associated with the viral DNA) from infected cells.

Specifications

Antigen Lac I
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Host Species Rabbit
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.