Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LACC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559081
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559081 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559081 Supplier Invitrogen™ Supplier No. PA559081
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FGLKLNSQKN CHQTLLKTLN AVQYHHAAKA KFLCIMCCSN ISYERDGEQD NCEIETSNGL SALLEEFEIV SCPSMAATLY TIKQKIDEKN LSSIKVI Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 59%.

This gene encodes an oxidoreductase that promotes fatty-acid oxidation, with concomitant inflammasome activation, mitochondrial and NADPH-oxidase-dependent reactive oxygen species production, and bactericidal activity of macrophages. The encoded protein forms a complex with fatty acid synthase on peroxisomes and is thought to be modulated by peroxisome proliferator-activated receptor signaling events. Naturally occurring mutations in this gene are associated with inflammatory bowel disease, Behcet's disease, leprosy, ulcerative colitis, early-onset Crohn's disease, and systemic juvenile idiopathic arthritis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LACC1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LACC1
Gene Accession No. Q8IV20
Gene Alias 9030625A04Rik; C12H13orf31; C13orf31; FAMIN; Fatty acid metabolism-immunity nexus; LACC1; laccase (multicopper oxidoreductase) domain containing 1; laccase domain containing 1; laccase domain-containing protein 1; purine nucleoside phosphorylase LACC1; laccase domain-containing protein 1; RGD1310185; si:ch211-187f22.1
Gene Symbols LACC1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LACC1. Recombinant protein control fragment (Product #RP-98003).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 144811
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.