Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LACE1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556369
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556369 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556369 Supplier Invitrogen™ Supplier No. PA556369
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NGLQRANFVP FIAVLKEYCN TVQLDSGIDY RKRELPAAGK LYYLTSEADV EAVMDKLFDE LAQKQNDLT Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 27%.

LACE1 encodes a protein with possible ATPase function. The protein contains a P-loop motif and a five-domain structure that is conserved in fly, yeast, and bacteria. Two conserved estrogen receptor binding sites are located within 2.5 kb of this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LACE1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene AFG1L
Gene Accession No. Q3V384, Q8WV93
Gene Alias AFG1; Afg1l; AFG1-like ATPase; ATPase family gene 1 homolog; c222389; CG8520; CG8520 gene product; Lace1; lactation elevated 1; lactation elevated protein 1; Protein AFG1 homolog
Gene Symbols AFG1L
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LACE1. Recombinant protein control fragment (Product #RP-94847).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 215951, 246269
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.