Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LACE1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598623
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598623 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598623 Supplier Invitrogen™ Supplier No. PA598623
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LACE1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

LACE1 encodes a protein with possible ATPase function. The protein contains a P-loop motif and a five-domain structure that is conserved in fly, yeast, and bacteria. Two conserved estrogen receptor binding sites are located within 2.5 kb of this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LACE1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene AFG1L
Gene Accession No. Q32PX9, Q8WV93
Gene Alias AFG1; Afg1l; AFG1-like ATPase; ATPase family gene 1 homolog; c222389; CG8520; CG8520 gene product; Lace1; lactation elevated 1; lactation elevated protein 1; Protein AFG1 homolog
Gene Symbols AFG1L
Host Species Rabbit
Immunogen Recombinant Human AFG1-like ATPase protein (14-313AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 246269, 502479
Target Species Human, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.